CAS RN 1373920-70-7
IUPAC Name:
3-bromo-4-methoxy-5-(trifluoromethyl)aniline
Synonyms:
3-bromo-4-methoxy-5-(trifluoromethyl)aniline
Molecular Weight (g/mol):
270.05
Molecular Formula:
C8H7BrF3NO
InChi Key:
ZGHWRZMMLVKQQA-UHFFFAOYSA-N
SMILES:
COC1=C(Br)C=C(N)C=C1C(F)(F)F
1
–
1
of
1
results
1
3-Bromo-4-methoxy-5-(trifluoromethyl)aniline, 97%, Thermo Scientific™
CAS: 1373920-70-7 Molecular Formula: C8H7BrF3NO Molecular Weight (g/mol): 270.049 MDL Number: MFCD22201052 InChI Key: ZGHWRZMMLVKQQA-UHFFFAOYSA-N Synonym: 3-bromo-4-methoxy-5-trifluoromethyl aniline PubChem CID: 86277652 IUPAC Name: 3-bromo-4-methoxy-5-(trifluoromethyl)aniline SMILES: COC1=C(C=C(C=C1C(F)(F)F)N)Br