accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNNFFFH2N

CAS RN 1372711-01-7

IUPAC Name: 2-(1-methylhydrazin-1-yl)-4-(trifluoromethyl)pyrimidine
Synonyms: 2-(1-methylhydrazin-1-yl)-4-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 192.15
Molecular Formula: C6H7F3N4
InChi Key: KSHPFMMNRGWDRW-UHFFFAOYSA-N
SMILES: CN(N)C1=NC=CC(=N1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results