accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13718-59-7

BaOSeOO

CAS RN 13718-59-7

IUPAC Name: barium(2+) selenite
Synonyms: barium(2+) selenite
Molecular Weight (g/mol): 264.3
Molecular Formula: BaO3Se
InChi Key: VQKDPABSEYUERS-UHFFFAOYSA-L
SMILES: [Ba++].[O-][Se]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results