accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 137-18-8

CH3OOCH3

CAS RN 137-18-8

IUPAC Name: 2,5-dimethylcyclohexa-2,5-diene-1,4-dione
Synonyms: 2,5-dimethylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: MYKLQMNSFPAPLZ-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C(C)=CC1=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results