CAS RN 13692-15-4
CAS RN 13692-15-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(2,4-Dichlorophenyl)-oxirane, TRC
CAS: 13692-15-4 Molecular Formula: C8H6OCl2 Molecular Weight (g/mol): 189.03 Synonym: Oxirane, 2-(2,4-dichlorophenyl)- (ACI),2-(2,4-Dichlorophenyl)oxirane (ACI),Benzene, 2,4-dichloro-1-(epoxyethyl)- (8CI),Oxirane, (2,4-dichlorophenyl)- (9CI),(2,4-Dichlorophenyl)oxirane IUPAC Name: 2-(2,4-dichlorophenyl)oxirane SMILES: Clc1ccc(C2CO2)c(Cl)c1