accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13686-66-3

H3CNCH3SiCH3H3C

CAS RN 13686-66-3

IUPAC Name: (diethylamino)dimethylsilyl
Synonyms: (diethylamino)dimethylsilyl
Molecular Weight (g/mol): 130.29
Molecular Formula: C6H16NSi
InChi Key: ADTGAVILDBXARD-UHFFFAOYSA-N
SMILES: CCN(CC)[Si](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results