accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13685-26-2

PClClCl

CAS RN 13685-26-2

IUPAC Name: chlorobis(4-chlorophenyl)phosphane
Synonyms: chlorobis(4-chlorophenyl)phosphane
Molecular Weight (g/mol): 289.52
Molecular Formula: C12H8Cl3P
InChi Key: UISLTICQIVPVGS-UHFFFAOYSA-N
SMILES: ClP(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)