accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 136849-72-4

IUPAC Name: ethyl cyano({[(dimethylamino)(dimethyliminiumyl)methoxy]imino})formate; tetrafluoroboranuide
Synonyms: ethyl cyano({[(dimethylamino)(dimethyliminio)methoxy]imino})formate tetrafluoroborate
Molecular Weight (g/mol): 328.07
Molecular Formula: C10H17BF4N4O3
InChi Key: FPQVGDGSRVMNMR-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCOC(=O)C(=NOC(N(C)C)=[N+](C)C)C#N
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results