accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClNOONOOOOH

CAS RN 136833-36-8

IUPAC Name: 5-chloro-2,4-dinitrobenzoic acid
Synonyms: 5-chloro-2,4-dinitrobenzoic acid
Molecular Weight (g/mol): 246.56
Molecular Formula: C7H3ClN2O6
InChi Key: VVQOEJCXWMGEJG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results