accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 136735-95-0

IUPAC Name: 1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethylphospholane
Synonyms: 1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethylphospholane
Molecular Weight (g/mol): 306.37
Molecular Formula: C18H28P2
InChi Key: AJNZWRKTWQLAJK-UHFFFAOYNA-N
SMILES: CC1CCC(C)P1C1=CC=CC=C1P1C(C)CCC1C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results