accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COPFFFFFFFFFFFFPFFFFFFFFFFFFOH3COH3COH3C(?)

CAS RN 1365531-85-6

IUPAC Name: bis[3,5-bis(trifluoromethyl)phenyl](2'-{bis[3,5-bis(trifluoromethyl)phenyl]phosphanyl}-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl)phosphane
Synonyms: bis[3,5-bis(trifluoromethyl)phenyl](2'-{bis[3,5-bis(trifluoromethyl)phenyl]phosphanyl}-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl)phosphane
Molecular Weight (g/mol): 1186.66
Molecular Formula: C48H28F24O4P2
InChi Key: JDKXXRZDVSJSNQ-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C(=C1)P(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=C(OC)C=C(OC)C=C1P(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results