accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNNH3CH3COOHOOH(E/Z)

CAS RN 136152-65-3

IUPAC Name: but-2-enedioic acid; dimethyl(2-{3-[1-(pyridin-2-yl)ethyl]-1H-inden-2-yl}ethyl)amine
Synonyms: butenedioic acid; dimethindene
Molecular Weight (g/mol): 408.5
Molecular Formula: C24H28N2O4
InChi Key: SWECWXGUJQLXJF-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CC(C1=C(CCN(C)C)CC2=CC=CC=C12)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results