accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 136099-65-5

IUPAC Name: (5-methyl-1-benzothiophen-2-yl)boronic acid
Synonyms: 5-methyl-1-benzothiophen-2-ylboronic acid
Molecular Weight (g/mol): 192.04
Molecular Formula: C9H9BO2S
InChi Key: DHNHZPQPQAINDI-UHFFFAOYSA-N
SMILES: CC1=CC=C2SC(=CC2=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results