accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13597-46-1

ZnOSeOO

CAS RN 13597-46-1

IUPAC Name: zinc(2+) selenite
Synonyms: zinc(2+) selenite
Molecular Weight (g/mol): 192.35
Molecular Formula: O3SeZn
InChi Key: PEUPCBAALXHYHP-UHFFFAOYSA-L
SMILES: [Zn++].[O-][Se]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results