accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13595-87-4

OOOOOOOOOWWBiBi

CAS RN 13595-87-4

IUPAC Name: dibismuth(3+) ditungsten nonaoxidandiide
Synonyms: dibismuth(3+) ditungsten nonaoxidandiide
Molecular Weight (g/mol): 929.64
Molecular Formula: Bi2O9W2
InChi Key: WLZFQTYMTGPFOM-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[W].[W].[Bi+3].[Bi+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results