accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 135911-02-3

H3COHNN(R)(R)O

CAS RN 135911-02-3

IUPAC Name: (3aR,7aR)-2-[2-(2-methoxyphenyl)ethanimidoyl]-7,7-diphenyl-octahydro-1H-isoindol-4-one
Synonyms: (3aR,7aR)-2-[2-(2-methoxyphenyl)ethanimidoyl]-7,7-diphenyl-hexahydroisoindol-4-one
Molecular Weight (g/mol): 438.57
Molecular Formula: C29H30N2O2
InChi Key: VWBOQFANCXZMAU-LOSJGSFVSA-N
SMILES: COC1=CC=CC=C1CC(=N)N1C[C@H]2[C@@H](C1)C(CCC2=O)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results