CAS RN 1357470-29-1
CAS RN 1357470-29-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ON123300, MedChemExpress
MedChemExpress ON123300, a strong and brain-penetrant multi-kinase inhibitor, inhibits CDK4 (IC50=3.9 nM), Ark5 (IC50=5 nM), PDGFRβ (IC50=26 nM), FGFR1 (IC50=26 nM), RET (IC50=9.2 nM), and FYN (IC50=11 nM). Single agent ON123300 causes a dose-dependent suppression of phosphorylation of Akt as well as activation of Erk in brain tumors. ON123300 inhibits CDK6 with an IC50 of 9.82 nM.