accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 135611-32-4

NNOOHHCl

CAS RN 135611-32-4

IUPAC Name: 4-[(1H-imidazol-1-yl)methyl]benzoic acid hydrochloride
Synonyms: 4-(imidazol-1-ylmethyl)benzoic acid hydrochloride
Molecular Weight (g/mol): 238.67
Molecular Formula: C11H11ClN2O2
InChi Key: MZTHHEXWAYFTBJ-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=CC=C(CN2C=CN=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results