accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 135564-61-3

CH3FClO

CAS RN 135564-61-3

IUPAC Name: 2-fluoro-5-methylbenzoyl chloride
Synonyms: 2-fluoro-5-methylbenzoyl chloride
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: WVRZOPWHCFDYSQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results