accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1355241-48-3

NHNSNH2BrBr

CAS RN 1355241-48-3

IUPAC Name: (2,6-dibromopyridin-3-yl)thiourea
Synonyms: 2,6-dibromopyridin-3-ylthiourea
Molecular Weight (g/mol): 311
Molecular Formula: C6H5Br2N3S
InChi Key: JYUVZMXPLAAKLL-UHFFFAOYSA-N
SMILES: NC(=S)NC1=C(Br)N=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results