accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1352999-98-4

OFFFFO

CAS RN 1352999-98-4

IUPAC Name: 3-fluoro-5-(trifluoromethoxy)benzaldehyde
Synonyms: 3-fluoro-5-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 208.11
Molecular Formula: C8H4F4O2
InChi Key: QIBNSINERFPXSF-UHFFFAOYSA-N
SMILES: FC1=CC(C=O)=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results