accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1352999-96-2

OFFFFNH2

CAS RN 1352999-96-2

IUPAC Name: 3-fluoro-5-(trifluoromethoxy)aniline
Synonyms: 3-fluoro-5-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 195.12
Molecular Formula: C7H5F4NO
InChi Key: NLCVCBKAVVNGQB-UHFFFAOYSA-N
SMILES: NC1=CC(OC(F)(F)F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results