accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OFFFFOHO

CAS RN 1352999-94-0

IUPAC Name: 2-[3-fluoro-5-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [3-fluoro-5-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 238.14
Molecular Formula: C9H6F4O3
InChi Key: LEUVGYUDBIAFFK-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(OC(F)(F)F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results