accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1352999-93-9

OFFFFN

CAS RN 1352999-93-9

IUPAC Name: 3-fluoro-5-(trifluoromethoxy)benzonitrile
Synonyms: 3-fluoro-5-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 205.11
Molecular Formula: C8H3F4NO
InChi Key: WACIMAFLAUYVCN-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(OC(F)(F)F)=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results