accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13517-24-3

H2OH2OH2OH2OH2OH2OH2OH2OH2ONaNaOSiOO

CAS RN 13517-24-3

IUPAC Name: disodium oxosilanebis(olate) nonahydrate
Synonyms: disodium oxosilanebis(olate) nonahydrate
Molecular Weight (g/mol): 284.2
Molecular Formula: H18Na2O12Si
InChi Key: PHIQPXBZDGYJOG-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Grade 
  • (1)
  • (4)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)