accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13517-23-2

H2OH2OH2OH2OH2OOPHOONaNa

CAS RN 13517-23-2

IUPAC Name: disodium pentahydrate phosphonate
Synonyms: disodium pentahydrate phosphonate
Molecular Weight (g/mol): 216.03
Molecular Formula: H11Na2O8P
InChi Key: VCNBZJAEINSDQB-UHFFFAOYSA-L
SMILES: O.O.O.O.O.[Na+].[Na+].[O-]P([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results