accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13517-10-7

IBII

CAS RN 13517-10-7

IUPAC Name: triiodoborane
Synonyms: boron triiodide
Molecular Weight (g/mol): 391.52
Molecular Formula: BI3
InChi Key: YMEKEHSRPZAOGO-UHFFFAOYSA-N
SMILES: IB(I)I
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results