CAS RN 1350622-33-1
CAS RN 1350622-33-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
kb-NB77-78, MedChemExpress
MedChemExpress kb-NB77-78 is an analogue of CID797718, but shows no PKD inhibitory activity.