accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13484-40-7

H3CONHNH2

CAS RN 13484-40-7

IUPAC Name: 4-(2-methoxyethyl)piperazine-1,4-diium
Synonyms: 4-(2-methoxyethyl)piperazine-1,4-diium
Molecular Weight (g/mol): 146.23
Molecular Formula: C7H18N2O
InChi Key: BMEMBBFDTYHTLH-UHFFFAOYSA-P
SMILES: COCC[NH+]1CC[NH2+]CC1
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)