accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1347815-30-8

OOHNNN

CAS RN 1347815-30-8

IUPAC Name: 2-(pyridin-4-yl)-3H,6H,11H-anthra[1,2-d]imidazole-6,11-dione
Synonyms: 2-(pyridin-4-yl)-3H-anthra[1,2-d]imidazole-6,11-dione
Molecular Weight (g/mol): 325.33
Molecular Formula: C20H11N3O2
InChi Key: YZNTXTFZKAVVRN-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=O)C2=C3N=C(NC3=CC=C12)C1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results