accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1347814-96-3

IUPAC Name: 5-chloro-2-phenylfuro[3,2-b]pyridine
Synonyms: 5-chloro-2-phenylfuro[3,2-b]pyridine
Molecular Weight (g/mol): 229.66
Molecular Formula: C13H8ClNO
InChi Key: XPTZPTVRNMXQEA-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC(=CC2=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results