accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1347814-83-8

IUPAC Name: 5-bromo-2-(4-methoxyphenyl)furo[2,3-b]pyridine
Synonyms: 5-bromo-2-(4-methoxyphenyl)furo[2,3-b]pyridine
Molecular Weight (g/mol): 304.14
Molecular Formula: C14H10BrNO2
InChi Key: WWILJVHMJKXSCO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC2=CC(Br)=CN=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results