accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13477-95-7

CNCNH2OH2OH2OH2ONi

CAS RN 13477-95-7

IUPAC Name: nickel(2+) bis(iminomethanide) tetrahydrate
Synonyms: nickel(2+) bis(cyanide) tetrahydrate
Molecular Weight (g/mol): 182.79
Molecular Formula: C2H8N2NiO4
InChi Key: VHHVAACTKRUWEK-UHFFFAOYSA-N
SMILES: O.O.O.O.[Ni++].[C-]#N.[C-]#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results