accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13470-38-7

YIII

CAS RN 13470-38-7

IUPAC Name: yttrium(3+) triiodide
Synonyms: yttrium(3+) triiodide
Molecular Weight (g/mol): 469.62
Molecular Formula: I3Y
InChi Key: LFWQXIMAKJCMJL-UHFFFAOYSA-K
SMILES: [Y+3].[I-].[I-].[I-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results