accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13470-26-3

VBrBrBr

CAS RN 13470-26-3

IUPAC Name: vanadium(3+) tribromide
Synonyms: vanadium(III) bromide
Molecular Weight (g/mol): 290.65
Molecular Formula: Br3V
InChi Key: ZOYIPGHJSALYPY-UHFFFAOYSA-K
SMILES: [V+3].[Br-].[Br-].[Br-]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results