accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13470-04-7

SrOMoOOO

CAS RN 13470-04-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 247.57
Molecular Formula: MoO4Sr
InChi Key: YZJCJRRZBDFNQA-UHFFFAOYSA-N
SMILES: [Sr++].[O-][Mo]([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)