CAS RN 1346606-26-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
all-trans 4-Keto Retinoic Acid-(9-methyl)-d3, TRC
CAS: 1346606-26-5 Molecular Formula: C20 2H3 H23 O3 Molecular Weight (g/mol): 317.44 Synonym: 4-Oxoretinoic-(9-methyl-d3) acid,4-Ketoretinoic acid (9-methyl-D3),4-Oxo-all-trans-retinoic-d3 acid,4-Oxotretinoin-d3,4-keto-Retinoic acid-d3,all-trans-4-Oxoretinoic acid-d3,(2E,4E,6E,8E)-3-Methyl-7-(methyl-d3)-9-(2,6,6-trimethyl-3-oxocyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid IUPAC Name: (2E,4E,6E,8E)-3-methyl-7-(trideuteriomethyl)-9-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid SMILES: [2H]C([2H])([2H])\C(=C/C=C/C(=C/C(=O)O)/C)\C=C\C1=C(C)C(=O)CCC1(C)C