CAS RN 13455-36-2
CAS RN 13455-36-2
IUPAC Name:
tris(λ²-cobalt(2+)) diphosphate
Synonyms:
cobalt phosphate
Molecular Weight (g/mol):
366.74
Molecular Formula:
Co3O8P2
InChi Key:
ZBDSFTZNNQNSQM-UHFFFAOYSA-H
SMILES:
[Co++].[Co++].[Co++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
1
–
1
of
1
results
1
Cobalt(II) phosphate, anhydrous, 98%
CAS: 13455-36-2 Molecular Formula: Co3O8P2 Molecular Weight (g/mol): 366.739 MDL Number: MFCD00016028 InChI Key: ZBDSFTZNNQNSQM-UHFFFAOYSA-H Synonym: cobalt phosphate,cobaltous phosphate,cobalt ii phosphate,tricobalt bis orthophosphate,unii-0b5m38t47h,cobalt ii phosphate, anhydrous,phosphoric acid, cobalt 2+ salt 2:3,3co.2po4,cobalt ii phosphate octahydrate,phosphoric acid,cobalt 2+ salt 2:3 PubChem CID: 61615 IUPAC Name: cobalt(2+);diphosphate SMILES: [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]