accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13455-25-9

CoOCrOOO

CAS RN 13455-25-9

IUPAC Name: λ²-cobalt(2+) dioxochromiumbis(olate)
Synonyms: λ²-cobalt(2+) chromate(2-)
Molecular Weight (g/mol): 174.93
Molecular Formula: CoCrO4
InChi Key: XTUHPOUJWWTMNC-UHFFFAOYSA-N
SMILES: [Co++].[O-][Cr]([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results