accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13454-70-1

MoCeO3212

CAS RN 13454-70-1

IUPAC Name: trimolybdenum(6+) dicerium(3+) dodecaoxidandiide
Synonyms: trimolybdenum(6+) dicerium(3+) dodecaoxidandiide
Molecular Weight (g/mol): 760.07
Molecular Formula: Ce2Mo3O12
InChi Key: BPOYBXJKAISYKA-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo+6].[Mo+6].[Mo+6].[Ce+3].[Ce+3]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results