accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13453-33-3

H2OClClClTl

CAS RN 13453-33-3

IUPAC Name: thallium(3+) hydrate trichloride
Synonyms: thallium(3+) hydrate trichloride
Molecular Weight (g/mol): 328.75
Molecular Formula: Cl3H2OTl
InChi Key: SWGLTBCHNXCQPK-UHFFFAOYSA-K
SMILES: O.[Cl-].[Cl-].[Cl-].[Tl+3]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results