accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13444-98-9

H2OH2OOSOOOPd

CAS RN 13444-98-9

IUPAC Name: palladium(2+) dihydrate sulfate
Synonyms: palladium(2+) dihydrate sulfate
Molecular Weight (g/mol): 238.51
Molecular Formula: H4O6PdS
InChi Key: TWIRRPLUAGEFNJ-UHFFFAOYSA-L
SMILES: O.O.[Pd++].[O-]S([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)