accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 134419-59-3

H3CSOOOO

CAS RN 134419-59-3

IUPAC Name: oxan-4-yl methanesulfonate
Synonyms: oxan-4-yl methanesulfonate
Molecular Weight (g/mol): 180.22
Molecular Formula: C6H12O4S
InChi Key: GSEZHCLWHDZJAB-UHFFFAOYSA-N
SMILES: CS(=O)(=O)OC1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results