accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 134412-18-3

IUPAC Name: 4'-(4-ethylcyclohexyl)-3,4-difluoro-1,1'-biphenyl
Synonyms: 4'-(4-ethylcyclohexyl)-3,4-difluoro-1,1'-biphenyl
Molecular Weight (g/mol): 300.39
Molecular Formula: C20H22F2
InChi Key: TYZNCUASDJPXMP-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results