accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NNHSNH

CAS RN 13431-41-9

IUPAC Name: 3-amino-1-benzylthiourea
Synonyms: 3-amino-1-benzylthiourea
Molecular Weight (g/mol): 181.26
Molecular Formula: C8H11N3S
InChi Key: ZTRUHAVBRPABTK-UHFFFAOYSA-N
SMILES: NNC(=S)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results