accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 134108-92-2

H3COHO

CAS RN 134108-92-2

IUPAC Name: 2-methoxycyclohexan-1-ol
Synonyms: 2-methoxycyclohexan-1-ol
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: DCQQZLGQRIVCNH-UHFFFAOYNA-N
SMILES: COC1CCCCC1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results