accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 134052-73-6

OOHH2NOHO

CAS RN 134052-73-6

IUPAC Name: 4-[amino(carboxy)methyl]benzoic acid
Synonyms: 4-carboxyphenylglycine (S)-4-carboxyphenylglycine 4-cpg 4-carboxylphenylglycine
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: VTMJKPGFERYGJF-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results