accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNNNNH2Br

CAS RN 1337532-51-0

IUPAC Name: 5-bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
Synonyms: 5-bromo-7-methylpyrrolo[2,3-d]pyrimidin-4-amine
Molecular Weight (g/mol): 227.07
Molecular Formula: C7H7BrN4
InChi Key: SCHJLAFNBDGWJN-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C2=C(N)N=CN=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results