accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3OON

CAS RN 13361-30-3

IUPAC Name: propan-2-yl 2-cyanoacetate
Synonyms: isopropyl 2-cyanoacetate
Molecular Weight (g/mol): 127.14
Molecular Formula: C6H9NO2
InChi Key: BESQLCCRQYTQQI-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results