accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13360-63-9

H3CHNCH3

CAS RN 13360-63-9

IUPAC Name: butyl(ethyl)amine
Synonyms: butethamine 丁胺卡因
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: QHCCDDQKNUYGNC-UHFFFAOYSA-N
SMILES: CCCCNCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results